Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPDL3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009568.2 | NP_001009568.1 | 5 | 893 | ||
NM_001304579.1 | NP_001291508.1 | 5 | 964 | |||
NM_014474.3 | NP_055289.2 | 5 | 893 | |||
XM_011541259.2 | XP_011539561.1 | 6 | 983 | |||
XM_011541260.2 | XP_011539562.1 | 6 | 1618 | |||
GenBank mRNA | AK293489.1 | - | 4 | 595 | ||
AK293566.1 | - | 5 | 890 | |||
AK301349.1 | - | 5 | 848 | |||
AK316500.1 | - | 5 | 819 | |||
BC014444.1 | - | 5 | 810 | 4865268 | ||
Y08134.1 | - | 5 | 785 |