Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014494.2 | NP_055309.2 | 22 | 5348 | ||
XM_005255254.3 | XP_005255311.1 | 21 | 5191 | |||
XM_005255257.4 | XP_005255314.1 | 21 | 5315 | |||
XM_017023143.1 | XP_016878632.1 | 21 | 5292 | |||
XM_017023144.1 | XP_016878633.1 | 20 | 5259 | |||
XM_017023145.1 | XP_016878634.1 | 21 | 5225 | |||
XM_017023146.1 | XP_016878635.1 | 21 | 5357 | |||
XM_017023147.1 | XP_016878636.1 | 21 | 5385 | |||
XM_017023148.1 | XP_016878637.1 | 20 | 5112 | |||
XM_017023149.1 | XP_016878638.1 | 19 | 5109 | |||
XM_017023150.1 | XP_016878639.1 | 20 | 5074 | |||
XM_017023151.1 | XP_016878640.1 | 19 | 4963 | |||
XM_017023152.1 | XP_016878641.1 | 20 | 4864 | |||
XM_017023153.1 | XP_016878642.1 | 21 | 5315 | |||
XM_017023154.1 | XP_016878643.1 | 19 | 5206 | |||
GenBank mRNA | AB040893.1 | - | 17 | 3548 | ||
AJ492221.1 | - | 21 | 5234 | |||
AK001773.1 | - | 5 | 673 | |||
AK025696.1 | - | 15 | 1905 | |||
AL833757.1 | - | 16 | 2509 | |||
AY035864.1 | - | 19 | 4882 | |||
BC024324.1 | - | 5 | 694 |