Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014505.5 | NP_055320.4 | 3 | 1070 | ||
GenBank mRNA | AF160967.1 | - | 3 | 954 | ||
AF207992.1 | - | 3 | 617 | |||
AF215891.1 | - | 3 | 672 | |||
BC042446.2 | - | 3 | 779 | 3873518 | ||
BC050621.1 | - | 3 | 711 | 5552807 | ||
BU556960.1 | - | 1 | 558 |