Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317893.1 | NP_001304822.1 | 2 | 377 | ||
NM_001317894.1 | NP_001304823.1 | 2 | 377 | |||
NM_014506.2 | NP_055321.1 | 2 | 377 | |||
NR_133948.1 | 2 | 377 | ||||
NR_134113.1 | 2 | 377 | ||||
XM_005251927.3 | XP_005251984.1 | 2 | 372 | |||
GenBank mRNA | AF007872.1 | - | 1 | 98 | ||
AF317129.1 | - | 2 | 367 | |||
AI568476.1 | - | 1 | 75 | |||
BC015578.2 | - | 2 | 322 | 4589479 | ||
BE315222.1 | - | 2 | 251 | |||
BP366330.1 | - | 2 | 377 |