Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOBEC3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014508.2 | NP_055323.2 | 4 | 611 | ||
GenBank mRNA | AB208979.1 | - | 4 | 576 | ||
AK094570.1 | - | 4 | 1925 | |||
AK313272.1 | - | 4 | 615 | |||
BC011739.2 | - | 4 | 611 | 4331363 | ||
BC021080.1 | - | 4 | 598 | |||
CR456394.1 | - | 4 | 589 | |||
KC329528.1 | - | 4 | 508 |