Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301849.1 | NP_001288778.1 | 10 | 1317 | ||
NM_015705.5 | NP_056520.2 | 11 | 1462 | |||
XM_005261572.2 | XP_005261629.1 | 11 | 1489 | |||
XM_005261573.3 | XP_005261630.1 | 11 | 1494 | |||
XM_005261574.4 | XP_005261631.1 | 11 | 1494 | |||
XM_005261575.1 | XP_005261632.1 | 10 | 1357 | |||
XM_011530158.1 | XP_011528460.1 | 10 | 1313 | |||
XM_011530159.1 | XP_011528461.1 | 11 | 1377 | |||
XM_011530160.1 | XP_011528462.1 | 10 | 1294 | |||
XM_011530161.1 | XP_011528463.1 | 11 | 1486 | |||
XM_011530162.2 | XP_011528464.1 | 11 | 1372 | |||
XM_017028775.1 | XP_016884264.1 | 10 | 1281 | |||
XM_017028776.1 | XP_016884265.1 | 10 | 1350 | |||
XM_017028777.1 | XP_016884266.1 | 10 | 1350 | |||
XM_017028778.1 | XP_016884267.1 | 9 | 1213 | |||
XM_017028779.1 | XP_016884268.1 | 10 | 1403 | |||
XM_017028780.1 | XP_016884269.1 | 10 | 1392 | |||
GenBank mRNA | AB196956.1 | - | 9 | 1231 | ||
AB275763.1 | - | 10 | 1335 | |||
AB449880.1 | - | 9 | 1224 | |||
AK000006.1 | - | 6 | 1556 | |||
AK295469.1 | - | 11 | 1408 | |||
AK297599.1 | - | 10 | 1310 | |||
AK301042.1 | - | 10 | 1230 | |||
AY308849.1 | - | 11 | 1397 | |||
BC008078.2 | - | 11 | 1388 | 3027777 | ||
BC127850.1 | - | 4 | 538 | |||
CR456441.1 | - | 10 | 1260 |