Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145076.2 | NP_001138548.1 | 14 | 1842 | ||
NM_019063.4 | NP_061936.2 | 15 | 2016 | |||
XM_005264267.1 | XP_005264324.1 | 16 | 2029 | |||
XM_005264268.3 | XP_005264325.1 | 16 | 2029 | |||
XM_006711991.1 | XP_006712054.1 | 15 | 1855 | |||
XM_006711992.2 | XP_006712055.1 | 17 | 2150 | |||
GenBank mRNA | AF177377.2 | - | 15 | 1978 | ||
AK056880.1 | - | 4 | 518 | |||
AK295778.1 | - | 2 | 241 | |||
AK314702.1 | - | 15 | 1990 | |||
AL832392.1 | - | 2 | 237 | |||
BC140845.1 | - | 15 | 1939 | 9021713 | ||
BC146799.1 | - | 14 | 1816 | 8860304 | ||
BX647731.1 | - | 6 | 1079 |