Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145076.2 | NP_001138548.1 | 22 | 2891 | ||
NM_019063.4 | NP_061936.2 | 23 | 3065 | |||
XM_005264267.1 | XP_005264324.1 | 24 | 3078 | |||
XM_006711991.1 | XP_006712054.1 | 23 | 2904 | |||
XM_006711992.2 | XP_006712055.1 | 25 | 3199 | |||
GenBank mRNA | AF177377.2 | - | 23 | 3027 | ||
AK001804.1 | - | 491 | ||||
AK056880.1 | - | 12 | 1567 | |||
AK295778.1 | - | 8 | 1103 | |||
AK314702.1 | - | 23 | 3039 | |||
AL832392.1 | - | 9 | 1218 | |||
BC023522.2 | - | 2 | 455 | |||
BC140845.1 | - | 23 | 2988 | 9021713 | ||
BC146799.1 | - | 22 | 2865 | 8860304 |