Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145044.1 | NP_001138516.1 | 6 | 1398 | ||
NM_013272.3 | NP_037404.2 | 6 | 1398 | |||
NR_135775.1 | 6 | 1128 | ||||
XM_005254889.1 | XP_005254946.1 | 6 | 1287 | |||
XM_005254891.2 | XP_005254948.1 | 6 | 1117 | |||
XM_011521456.2 | XP_011519758.1 | 7 | 1198 | |||
GenBank mRNA | AB031050.2 | - | 6 | 1390 | ||
AF187816.1 | - | 6 | 1197 | |||
AF205074.1 | - | 6 | 1386 | |||
AK057031.1 | - | 4 | 745 | |||
AK097797.1 | - | 7 | 1198 | |||
AK290872.1 | - | 6 | 1236 | |||
AK308463.1 | - | 5 | 633 | |||
AK310306.1 | - | 6 | 1093 | |||
BC000585.2 | - | 6 | 1250 | 3163826 |