Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145044.1 | NP_001138516.1 | 9 | 1901 | ||
NM_013272.3 | NP_037404.2 | 9 | 1901 | |||
NR_135775.1 | 9 | 1631 | ||||
XM_005254889.1 | XP_005254946.1 | 9 | 1790 | |||
XM_005254891.2 | XP_005254948.1 | 9 | 1620 | |||
XM_011521456.2 | XP_011519758.1 | 10 | 1701 | |||
GenBank mRNA | AB031050.2 | - | 9 | 1893 | ||
AF187816.1 | - | 9 | 1700 | |||
AF205074.1 | - | 9 | 1889 | |||
AK057031.1 | - | 7 | 1248 | |||
AK097797.1 | - | 10 | 1701 | |||
AK290872.1 | - | 9 | 1739 | |||
AK308463.1 | - | 8 | 1136 | |||
AK310306.1 | - | 9 | 1596 | |||
BC000585.2 | - | 9 | 1753 | 3163826 |