Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145044.1 | NP_001138516.1 | 2 | 720 | ||
NM_013272.3 | NP_037404.2 | 2 | 720 | |||
NR_135775.1 | 2 | 450 | ||||
XM_005254889.1 | XP_005254946.1 | 2 | 609 | |||
XM_005254891.2 | XP_005254948.1 | 2 | 439 | |||
XM_011521456.2 | XP_011519758.1 | 3 | 520 | |||
GenBank mRNA | AB031050.2 | - | 2 | 712 | ||
AF187816.1 | - | 2 | 519 | |||
AF205074.1 | - | 2 | 708 | |||
AK097797.1 | - | 3 | 520 | |||
AK290872.1 | - | 2 | 558 | |||
AK310306.1 | - | 2 | 415 | |||
BC000585.2 | - | 2 | 572 | 3163826 | ||
HY004505.1 | - | 2 | 453 |