Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014008.4 | NP_054727.1 | 2 | 233 | ||
XM_005272599.3 | XP_005272656.1 | 2 | 165 | |||
GenBank mRNA | AJ005890.1 | - | 2 | 149 | ||
AK291976.1 | - | 2 | 194 | |||
AK296911.1 | - | 2 | 204 | |||
AK298264.1 | - | 2 | 228 | |||
AK316375.1 | - | 2 | 179 | |||
BC000972.2 | - | 2 | 180 | 3449051 | ||
BC011675.2 | - | 2 | 64 | 4299954 | ||
BG912845.1 | - | 2 | 219 | |||
BM146102.1 | - | 2 | 234 | |||
DC340647.1 | - | 2 | 230 |