Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014012.5 | NP_054731.2 | 5 | 944 | ||
XM_005260404.1 | XP_005260461.1 | 5 | 968 | |||
XM_011528795.1 | XP_011527097.1 | 5 | 962 | |||
XM_017027833.1 | XP_016883322.1 | 5 | 802 | |||
GenBank mRNA | AF084465.1 | - | 4 | 718 | ||
AF152863.1 | - | 5 | 926 | |||
AK313060.1 | - | 5 | 944 | |||
BC039813.1 | - | 5 | 930 | 5220518 | ||
BT020078.1 | - | 4 | 646 | |||
BT020079.1 | - | 4 | 646 |