Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145264.1 | NP_001138736.1 | 1, 2 | 230 | ||
NM_014017.3 | NP_054736.1 | 1, 2 | 230 | |||
GenBank mRNA | AF070659.1 | - | 1, 2 | 142 | ||
AK307086.1 | - | 1, 2 | 156 | |||
BC024190.2 | - | 1, 2 | 106 | 2819969 | ||
BM353842.1 | - | 1, 2 | 198 | |||
BU184679.1 | - | 1, 2 | 142 | |||
CA438137.1 | - | 3, 4 | 467 | |||
DB566869.1 | - | 12 |