Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017372.2 | NP_001017372.1 | 6 | 2239 | ||
NM_001317984.1 | NP_001304913.1 | 7 | 2063 | |||
NM_014031.4 | NP_054750.1 | 7 | 2014 | |||
GenBank mRNA | AF064254.1 | - | 6 | 1381 | ||
AK313456.1 | - | 7 | 1662 | |||
AL833117.1 | - | 6 | 1812 | |||
BC041945.1 | - | 6 | 2238 | 5301306 | ||
BX537383.1 | - | 7 | 1624 | |||
CR457129.1 | - | 6 | 1228 |