Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014035.2 | NP_054754.1 | 6 | 583 | ||
XM_005271972.4 | XP_005272029.1 | 6 | 472 | |||
XM_006714592.3 | XP_006714655.1 | 6 | 538 | |||
XM_011543349.2 | XP_011541651.1 | 7 | 603 | |||
XM_011543350.2 | XP_011541652.1 | 7 | 602 | |||
XM_017009394.1 | XP_016864883.1 | 6 | 408 | |||
GenBank mRNA | AF139461.1 | - | 6 | 472 | ||
AY044655.1 | - | 6 | 414 | |||
AY358098.1 | - | 6 | 432 | |||
BC010886.1 | - | 6 | 543 | 3538426 | ||
BC069012.1 | - | 6 | 530 | 4585781 | ||
BQ278046.1 | - | 6 | 555 |