Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAMDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316960.1 | NP_001303889.1 | 5 | 580 | ||
NM_001316961.1 | NP_001303890.1 | 5 | 500 | |||
NM_001316962.1 | NP_001303891.1 | 5 | 504 | |||
NM_024684.3 | NP_078960.1 | 4 | 462 | |||
XM_011544968.2 | XP_011543270.1 | 4 | 462 | |||
XM_011544969.2 | XP_011543271.1 | 4 | 455 | |||
GenBank mRNA | AI569731.1 | - | 1 | 86 | ||
AK024688.1 | - | 4 | 421 | |||
BC002752.2 | - | 4 | 414 | 3628061 | ||
BI791786.1 | - | 5 | 513 | |||
BM930331.1 | - | 5 | 449 | |||
CR457332.1 | - | 3 | 316 |