Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014041.3 | NP_054760.3 | 4 | 631 | ||
GenBank mRNA | AF092138.1 | - | 4 | 360 | ||
AK027426.1 | - | 4 | 360 | |||
AX775783.1 | - | 4 | 360 | |||
AX775785.1 | - | 5 | 1145 | |||
AX775787.1 | - | 4 | 340 | |||
AX775833.1 | - | 4 | 350 | |||
BC000884.1 | - | 4 | 340 | 3462307 | ||
BI826431.1 | - | 4 | 325 | |||
BQ423345.1 | - | 4 | 367 | |||
CA309210.1 | - | 1 | 466 | |||
L38852.1 | - | 4 | 350 |