Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014046.3 | NP_054765.1 | 2 | 298 | ||
GenBank mRNA | AF100761.1 | - | 2 | 153 | ||
AF151017.1 | - | 2 | 179 | |||
AK294086.1 | - | 2 | 160 | |||
AK311890.1 | - | 2 | 149 | |||
AL050361.1 | - | 2 | 157 | |||
BC005373.1 | - | 2 | 178 | 3933607 | ||
BG773408.1 | - | 2 | 289 | |||
BP369345.1 | - | 2 | 298 |