Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014049.4 | NP_054768.2 | 17 | 1915 | ||
NR_033426.1 | 17 | 2091 | ||||
GenBank mRNA | AB209427.1 | - | 15 | 4919 | ||
AF078854.1 | - | 5 | 447 | |||
AF327351.1 | - | 17 | 1745 | |||
AK022568.1 | - | 16 | 1841 | |||
AK024012.1 | - | 17 | 1759 | |||
AK027186.1 | - | 6 | 1337 | |||
AK130311.1 | - | 17 | 1786 | |||
AK297952.1 | - | 17 | 1959 | |||
BC001817.2 | - | 10 | 879 | |||
BC007970.2 | - | 17 | 1792 | 4304209 | ||
BC013354.2 | - | 17 | 1769 | 3935925 |
Gene Symbol: KIAA1257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017006939.1 | XP_016862428.1 | 16 | 8285 | ||
XM_017006940.1 | XP_016862429.1 | 16 | 8281 | |||
XM_017006941.1 | XP_016862430.1 | 16 | 7858 | |||
XM_017006942.1 | XP_016862431.1 | 16 | 8252 | |||
XM_017006943.1 | XP_016862432.1 | 16 | 8010 | |||
XM_017006944.1 | XP_016862433.1 | 15 | 7830 | |||
XM_017006946.1 | XP_016862435.1 | 14 | 7441 | |||
XM_017006947.1 | XP_016862436.1 | 13 | 7325 | |||
XM_017006948.1 | XP_016862437.1 | 15 | 7502 | |||
XM_017006949.1 | XP_016862438.1 | 14 | 7449 | |||
XM_017006950.1 | XP_016862439.1 | 14 | 12936 | |||
XM_017006951.1 | XP_016862440.1 | 14 | 13041 | |||
XM_017006952.1 | XP_016862441.1 | 13 | 7219 | |||
XM_017006953.1 | XP_016862442.1 | 13 | 7225 |