Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143779.1 | NP_001137251.1 | 7, 8 | 830 | ||
NM_014055.3 | NP_054774.2 | 7, 8 | 1248 | |||
NM_031473.3 | NP_113661.2 | 7, 8 | 1248 | |||
XM_011538232.2 | XP_011536534.1 | 7, 8 | 776 | |||
XM_017019217.1 | XP_016874706.1 | 7, 8 | 940 | |||
XM_017019218.1 | XP_016874707.1 | 7, 8 | 1248 | |||
GenBank mRNA | AF139540.1 | - | 7, 8 | 788 | ||
AF250326.1 | - | 6, 7 | 741 | |||
AF332010.1 | - | 7, 8 | 1206 | |||
AK091549.1 | - | 6, 7 | 608 | |||
AK291043.1 | - | 7, 8 | 1150 | |||
BC004536.2 | - | 7, 8 | 1084 | 3608705 | ||
BC005396.1 | - | 7, 8 | 750 | |||
BC108257.1 | - | 6, 7 | 713 | 6156440 |