Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASTE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288950.1 | NP_001275879.1 | 4 | 1762 | ||
NM_014065.3 | NP_054784.2 | 4 | 1987 | |||
NR_110229.1 | 4 | 1987 | ||||
XM_017006266.1 | XP_016861755.1 | 4 | 1903 | |||
XM_017006267.1 | XP_016861756.1 | 4 | 2061 | |||
XM_017006268.1 | XP_016861757.1 | 4 | 1982 | |||
XM_017006269.1 | XP_016861758.1 | 4 | 1903 | |||
GenBank mRNA | AF113539.1 | - | 2 | 657 | ||
AK027258.1 | - | 2 | 1377 | |||
AK027806.1 | - | 4 | 1935 | |||
AK294156.1 | - | 4 | 1987 | |||
AK301388.1 | - | 5 | 1552 | |||
BC030274.1 | - | 2 | 607 | |||
BC069265.1 | - | 4 | 1700 | |||
BC103709.1 | - | 2 | 1078 | |||
BC110616.2 | - | 4 | 1700 | 40033986 |