Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASTE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288950.1 | NP_001275879.1 | 7 | 2331 | ||
NM_014065.3 | NP_054784.2 | 6 | 2481 | |||
NR_110229.1 | 7 | 2541 | ||||
XM_017006266.1 | XP_016861755.1 | 7 | 2457 | |||
XM_017006267.1 | XP_016861756.1 | 7 | 2615 | |||
XM_017006268.1 | XP_016861757.1 | 7 | 3498 | |||
XM_017006269.1 | XP_016861758.1 | 7 | 3419 | |||
GenBank mRNA | AF113539.1 | - | 4 | 1151 | ||
AK027258.1 | - | 4 | 1871 | |||
AK294156.1 | - | 7 | 2542 | |||
BC030274.1 | - | 4 | 1101 | |||
BC069265.1 | - | 7 | 2269 | |||
BC103709.1 | - | 4 | 1572 | |||
BC110616.2 | - | 6 | 2194 | 40033986 | ||
BM992874.1 | - | 1 | 252 |
Gene Symbol: ATP2C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833453.1 | - | 1 | 1135 |