Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014079.3 | NP_054798.1 | 2 | 638 | ||
XM_005247400.3 | XP_005247457.1 | 2 | 2355 | |||
XM_011512743.2 | XP_011511045.1 | 2 | 583 | |||
GenBank mRNA | AB029254.1 | - | 1 | 456 | ||
BC036733.1 | - | 2 | 650 | 5526657 |