Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014149.3 | NP_054868.3 | 9 | 1431 | ||
XM_011516083.2 | XP_011514385.1 | 9 | 1401 | |||
XM_011516086.2 | XP_011514388.1 | 9 | 1401 | |||
XM_017012070.1 | XP_016867559.1 | 9 | 1401 | |||
XM_017012071.1 | XP_016867560.1 | 6 | 836 | |||
XM_017012072.1 | XP_016867561.1 | 9 | 1413 | |||
GenBank mRNA | AK001606.1 | - | 9 | 1386 | ||
AK022935.1 | - | 9 | 1375 | |||
AK074466.1 | - | 2 | 191 | |||
AL136940.1 | - | 9 | 1282 | |||
BC009939.2 | - | 7 | 889 | |||
BC017246.2 | - | 9 | 1370 | 4890904 | ||
BC063394.1 | - | 9 | 1367 | |||
BX640957.1 | - | 9 | 2351 | |||
CR533561.1 | - | 8 | 1213 |