Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014153.3 | NP_054872.2 | 23 | 2977 | ||
XM_006720877.2 | XP_006720940.1 | 23 | 3013 | |||
XM_011522463.1 | XP_011520765.1 | 24 | 3162 | |||
XM_011522464.2 | XP_011520766.1 | 17 | 2770 | |||
XM_017023174.1 | XP_016878663.1 | 23 | 2994 | |||
XM_017023175.1 | XP_016878664.1 | 19 | 2607 | |||
GenBank mRNA | AF161540.1 | - | 4 | 1796 | ||
AK000889.1 | - | 4 | 780 | |||
AK001869.1 | - | 4 | 844 | |||
AK026827.1 | - | 10 | 1100 | |||
AY163807.1 | - | 22 | 3006 | |||
BC012575.1 | - | 4 | 613 | |||
BC046363.1 | - | 23 | 2870 | 5538820 | ||
BX647154.1 | - | 2 | 576 |