Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318817.1 | NP_001305746.1 | 4 | 522 | ||
NM_014161.4 | NP_054880.2 | 4 | 671 | |||
NR_134860.1 | 5 | 791 | ||||
NR_134861.1 | 4 | 748 | ||||
NR_134862.1 | 4 | 671 | ||||
NR_134863.1 | 4 | 655 | ||||
NR_134864.1 | 4 | 554 | ||||
NR_134865.1 | 4 | 522 | ||||
NR_134866.1 | 4 | 500 | ||||
GenBank mRNA | AF161556.1 | - | 4 | 649 | ||
AK311391.1 | - | 5 | 668 | |||
AL136633.1 | - | 4 | 621 | |||
AV756740.1 | - | 4 | 457 | |||
BC001623.1 | - | 4 | 642 | 3357333 | ||
BF797811.1 | - | 4 | 555 | |||
BG548185.1 | - | 4 | 502 | |||
BM924328.1 | - | 4 | 764 | |||
BU601065.1 | - | 4 | 613 | |||
CA777760.1 | - | 4 | 382 | |||
CB529016.1 | - | 1 | 334 | |||
CD675657.1 | - | 4 | 554 | |||
CR533491.1 | - | 4 | 529 |