Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293186.1 | NP_001280115.1 | 2 | 266 | ||
NM_001293187.1 | NP_001280116.1 | 2 | 180 | |||
NM_014168.3 | NP_054887.2 | 1 | 407 | |||
XM_011511023.1 | XP_011509325.1 | 2 | 207 | |||
GenBank mRNA | AF161482.1 | - | 2 | 155 | ||
AF201938.1 | - | 2 | 224 | |||
AL523293.3 | - | 1 | 389 | |||
BC000921.2 | - | 1 | 319 | 3445582 | ||
BC093014.1 | - | 2 | 162 | 4799279 | ||
CA337169.1 | - | 2 | 181 |