Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135861.2 | NP_001129333.1 | 2 | 865 | ||
NM_001287342.1 | NP_001274271.1 | 2 | 871 | |||
NM_001287343.1 | NP_001274272.1 | 2 | 865 | |||
NM_014172.5 | NP_054891.2 | 2 | 865 | |||
NR_109807.1 | 2 | 865 | ||||
NR_109808.1 | 2 | 848 | ||||
GenBank mRNA | AA460844.1 | - | 2 | 338 | ||
AF131857.1 | - | 2 | 589 | |||
AF161490.1 | - | 2 | 270 | |||
AF164795.1 | - | 2 | 258 | |||
AF285119.1 | - | 2 | 583 | |||
AF393504.1 | - | 2 | 250 | |||
AL136644.1 | - | 2 | 275 | |||
BC024648.1 | - | 2 | 271 | 4934238 | ||
BM979912.1 | - | 2 | 744 | |||
BP375594.1 | - | 2 | 269 | |||
BQ922335.1 | - | 2 | 280 | |||
CD242442.1 | - | 2 | 478 | |||
CD693554.1 | - | 2 | 310 | |||
CN334799.1 | - | 2 | 248 | |||
CR533481.1 | - | 2 | 250 | |||
EU794646.1 | - | 2 | 582 | |||
F29783.1 | - | 2 | 108 |