Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BABAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033549.2 | NP_001028721.1 | 7 | 804 | ||
NM_001288756.1 | NP_001275685.1 | 7 | 755 | |||
NM_001288757.1 | NP_001275686.1 | 4 | 579 | |||
NM_014173.3 | NP_054892.2 | 7 | 766 | |||
GenBank mRNA | AF161491.1 | - | 7 | 727 | ||
AK000578.1 | - | 7 | 681 | |||
AK025970.1 | - | 7 | 699 | |||
AK225047.1 | - | 7 | 688 | |||
AK301193.1 | - | 4 | 474 | |||
AK308590.1 | - | 6 | 707 | |||
AL136692.1 | - | 7 | 698 | |||
BC000788.2 | - | 7 | 665 | 3504488 | ||
BC006244.1 | - | 7 | 706 | 3947341 | ||
BC091491.1 | - | 7 | 712 | 30364624 | ||
CR533526.1 | - | 6 | 601 | |||
CX866662.1 | - | 4 | 452 |