Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014175.3 | NP_054894.1 | 2, 3 | 315 | ||
XM_017013362.1 | XP_016868851.1 | 2, 3 | 326 | |||
GenBank mRNA | AF161494.1 | - | 2, 3 | 307 | ||
AK312832.1 | - | 2, 3 | 279 | |||
AL136665.1 | - | 2, 3 | 285 | |||
BC000891.1 | - | 2, 3 | 315 | 3463430 | ||
CR533531.1 | - | 2, 3 | 252 |