Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304476.1 | NP_001291405.1 | 4, 5 | 862 | ||
NM_001304477.1 | NP_001291406.1 | 3, 4 | 367 | |||
NM_014178.7 | NP_054897.4 | 3, 4 | 1009 | |||
XM_006720121.3 | XP_006720184.1 | 3, 4 | 372 | |||
XM_011536669.2 | XP_011534971.1 | 2, 3 | 404 | |||
XM_017021231.1 | XP_016876720.1 | 3, 4 | 741 | |||
XM_017021232.1 | XP_016876721.1 | 4, 5 | 3337 | |||
XM_017021233.1 | XP_016876722.1 | 4, 5 | 689 | |||
XM_017021234.1 | XP_016876723.1 | 4, 5 | 462 | |||
XM_017021235.1 | XP_016876724.1 | 3, 4 | 3250 | |||
XM_017021236.1 | XP_016876725.1 | 4, 5 | 666 | |||
XM_017021237.1 | XP_016876726.1 | 4, 5 | 495 | |||
XM_017021238.1 | XP_016876727.1 | 3, 4 | 433 | |||
XM_017021239.1 | XP_016876728.1 | 3, 4 | 431 | |||
XM_017021240.1 | XP_016876729.1 | 3, 4 | 353 | |||
XM_017021241.1 | XP_016876730.1 | 3, 4 | 970 | |||
XM_017021242.1 | XP_016876731.1 | 4, 5 | 435 | |||
GenBank mRNA | AF391153.1 | - | 2, 3 | 333 | ||
AK096957.1 | - | 5, 6 | 592 | |||
AK309203.1 | - | 3, 4 | 367 | |||
AL834346.1 | - | 3, 4 | 347 | |||
BC009499.2 | - | 4, 5 | 636 | 3955370 | ||
BC067278.1 | - | 3, 4 | 1009 | 6178786 |