Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277197.1 | NP_001264126.1 | 1 | 153 | ||
NM_001277198.1 | NP_001264127.1 | 1 | 153 | |||
NM_001277199.1 | NP_001264128.1 | 1 | 153 | |||
NM_001277200.1 | NP_001264129.1 | 1 | 153 | |||
NM_014184.3 | NP_054903.1 | 1 | 153 | |||
NR_102347.1 | 1 | 153 | ||||
GenBank mRNA | AF161512.1 | - | 1 | 72 | ||
AK024569.1 | - | 68 | ||||
AK289973.1 | - | 1 | 87 | |||
AK312064.1 | - | 1 | 78 | |||
AL136930.1 | - | 1 | 93 | |||
BC000573.1 | - | 1 | 68 | 3163724 | ||
BI752106.1 | - | 1 | 69 | |||
BQ051754.1 | - | 1 | 66 | |||
BX649076.1 | - | 1 | 57 | |||
HY018269.1 | - | 1 | 69 | |||
HY145928.1 | - | 1 | 78 |