Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013254.3 | NP_037386.1 | 9 | 1320 | ||
XM_005268809.1 | XP_005268866.1 | 9 | 1398 | |||
XM_005268810.1 | XP_005268867.1 | 9 | 1380 | |||
GenBank mRNA | AF174536.1 | - | 8 | 1161 | ||
AF191838.1 | - | 9 | 1223 | |||
AK002192.1 | - | 1 | 146 | |||
AK291039.1 | - | 9 | 1265 | |||
AK303683.1 | - | 4 | 537 |