Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014070.2 | NP_054789.2 | 2 | 206 | ||
GenBank mRNA | AB031481.1 | - | 2 | 206 | ||
AY358438.1 | - | 2 | 210 |