Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314029.1 | NP_001300958.1 | 3 | 329 | ||
NM_014143.3 | NP_054862.1 | 3 | 329 | |||
NR_052005.1 | 3 | 329 | ||||
GenBank mRNA | AF177937.1 | - | 2 | 221 | ||
AF233516.1 | - | 3 | 273 | |||
AK001894.1 | - | 1 | 36 | |||
AK300470.1 | - | 3 | 290 | |||
AK314567.1 | - | 3 | 290 | |||
AY254342.1 | - | 2 | 221 | |||
AY714881.1 | - | 2 | 221 | |||
BC069381.1 | - | 3 | 271 | 7262208 | ||
BC074984.2 | - | 2 | 241 | 30915301 | ||
BC113734.1 | - | 2 | 242 | 8322786 | ||
BC113736.1 | - | 2 | 242 | 8322788 | ||
DB160805.1 | - | 3 | 260 | |||
DQ836393.1 | - | 3 | 334 |