Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267706.1 | NP_001254635.1 | 5, 6 | 625 | ||
NM_014143.3 | NP_054862.1 | 6, 7 | 967 | |||
NR_052005.1 | 7, 8 | 794 | ||||
GenBank mRNA | AF177937.1 | - | 5, 6 | 859 | ||
AF233516.1 | - | 6, 7 | 911 | |||
AK001894.1 | - | 4, 5 | 674 | |||
AK314567.1 | - | 6, 7 | 928 | |||
AY254342.1 | - | 5, 6 | 859 | |||
AY291313.1 | - | 4, 5 | 517 | |||
BC069381.1 | - | 6, 7 | 909 | 7262208 | ||
BC074984.2 | - | 5, 6 | 879 | 30915301 | ||
BC113734.1 | - | 5, 6 | 880 | 8322786 | ||
BC113736.1 | - | 5, 6 | 880 | 8322788 | ||
DQ286582.1 | - | 4, 5 | 517 |