Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114094.1 | NP_001107566.1 | 12, 13 | 1037 | ||
NM_001258440.1 | NP_001245369.1 | 13, 14 | 1046 | |||
NM_001258441.1 | NP_001245370.1 | 12, 13 | 977 | |||
NM_001258442.1 | NP_001245371.1 | 10, 11 | 731 | |||
NM_013314.3 | NP_037446.1 | 13, 14 | 1106 | |||
NR_047680.1 | 12, 13 | 870 | ||||
NR_047681.1 | 11, 12 | 818 | ||||
NR_047682.1 | 11, 12 | 813 | ||||
NR_047683.1 | 11, 12 | 907 | ||||
XM_011539728.2 | XP_011538030.1 | 13, 14 | 983 | |||
XM_017016159.1 | XP_016871648.1 | 12, 13 | 915 | |||
GenBank mRNA | AF068180.1 | - | 13, 14 | 1081 | ||
AF068181.1 | - | 12, 13 | 1012 | |||
AK027779.1 | - | 13, 14 | 1041 | |||
AK225546.1 | - | 12, 13 | 1020 | |||
AM180327.1 | - | 10, 11 | 613 | |||
AM180332.1 | - | 12, 13 | 859 | |||
AM180333.1 | - | 12, 13 | 859 | |||
AM180334.1 | - | 12, 13 | 859 | |||
AM180335.1 | - | 9, 10 | 497 | |||
AM180336.1 | - | 13, 14 | 928 | |||
AM180337.1 | - | 13, 14 | 928 | |||
AM180338.1 | - | 11, 12 | 700 | |||
AM180339.1 | - | 11, 12 | 695 | |||
AM180340.1 | - | 11, 12 | 789 | |||
AM180342.1 | - | 12, 13 | 878 | |||
AM180343.1 | - | 12, 13 | 878 | |||
AM180344.1 | - | 10, 11 | 768 | |||
AM180345.1 | - | 10, 11 | 739 | |||
AM180346.1 | - | 10, 11 | 795 | |||
BC018906.2 | - | 13, 14 | 1020 | 4053894 | ||
BC110298.1 | - | 12, 13 | 862 | |||
BF975580.1 | - | 1, 2 | 37 |