Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321792.1 | NP_001308721.1 | 5, 6 | 655 | ||
NM_013342.3 | NP_037474.1 | 5, 6 | 1049 | |||
XM_005278261.1 | XP_005278318.1 | 4, 5 | 525 | |||
GenBank mRNA | AF052052.1 | - | 6, 7 | 922 | ||
BC004281.1 | - | 5, 6 | 681 | 3616596 | ||
BC007776.2 | - | 5, 6 | 667 | 4301790 | ||
BQ027375.1 | - | 1, 2 | 171 |