Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079514.2 | NP_001072982.1 | 15 | 3155 | ||
NM_001288656.1 | NP_001275585.1 | 14 | 2589 | |||
XM_005255277.3 | XP_005255334.1 | 15 | 3340 | |||
XM_005255278.3 | XP_005255335.1 | 15 | 4304 | |||
XM_005255279.3 | XP_005255336.1 | 14 | 3193 | |||
XM_011522465.1 | XP_011520767.1 | 15 | 3340 | |||
XM_011522466.2 | XP_011520768.1 | 10 | 1981 | |||
XM_017023189.1 | XP_016878678.1 | 15 | 3340 | |||
XM_017023190.1 | XP_016878679.1 | 14 | 3193 | |||
XM_017023191.1 | XP_016878680.1 | 8 | 1713 | |||
GenBank mRNA | AF108460.1 | - | 15 | 2650 | ||
AF108461.1 | - | 14 | 3237 | |||
AK300131.1 | - | 14 | 2589 |