Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142364.1 | NP_001135836.1 | 6, 7 | 605 | ||
NM_013338.4 | NP_037470.1 | 7, 8 | 695 | |||
XM_017020552.1 | XP_016876041.1 | 6, 7 | 633 | |||
GenBank mRNA | AF088028.1 | - | 2, 3 | 133 | ||
AF102850.1 | - | 7, 8 | 703 | |||
AF161498.1 | - | 7, 8 | 644 | |||
AF183413.1 | - | 7, 8 | 654 | |||
AK299085.1 | - | 6, 7 | 581 | |||
BC012531.1 | - | 7, 8 | 647 | 4273297 | ||
BU739112.1 | - | 3, 4 | 529 |