Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM216A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013300.2 | NP_037432.2 | 1, 2 | 598 | ||
GenBank mRNA | AK225731.1 | - | 1, 2 | 589 | ||
AK296280.1 | - | 1, 2 | 610 | |||
AK308802.1 | - | 1, 2 | 211 | |||
AK309603.1 | - | 1, 2 | 244 | |||
BC020967.1 | - | 1, 2 | 414 | 3848037 | ||
CR457001.1 | - | 1, 2 | 149 | |||
U79274.1 | - | 1, 2 | 565 |