Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307986.1 | NP_001294915.1 | 6 | 1250 | ||
NM_001307987.1 | NP_001294916.1 | 4 | 1049 | |||
NM_013305.5 | NP_037437.2 | 5 | 1142 | |||
XM_017025715.1 | XP_016881204.1 | 7 | 638 | |||
XM_017025716.1 | XP_016881205.1 | 5 | 816 | |||
GenBank mRNA | AK056270.1 | - | 5 | 1142 | ||
AK094273.1 | - | 4 | 655 | |||
AK130126.1 | - | 7 | 645 | |||
AK293608.1 | - | 6 | 890 | |||
AK298881.1 | - | 4 | 690 | |||
BC108910.1 | - | 5 | 692 | 40011279 | ||
BC108911.1 | - | 5 | 692 | 40011281 | ||
CR457037.1 | - | 5 | 481 | |||
U91641.1 | - | 5 | 528 |