Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013320.2 | NP_037452.1 | 7 | 1150 | ||
XM_017019241.1 | XP_016874730.1 | 7 | 1150 | |||
XM_017019242.1 | XP_016874731.1 | 7 | 1150 | |||
GenBank mRNA | AF117210.1 | - | 7 | 1059 | ||
AK223342.1 | - | 7 | 1077 | |||
AK313466.1 | - | 7 | 1080 | |||
BC006558.2 | - | 7 | 1066 | 2823984 | ||
BC033799.1 | - | 7 | 1052 | 5172046 |