Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013323.2 | NP_037455.2 | 3 | 364 | ||
NM_152244.1 | NP_689450.1 | 4 | 451 | |||
XM_005257260.3 | XP_005257317.1 | 3 | 131 | |||
XM_005257261.3 | XP_005257318.1 | 3 | 398 | |||
XM_005257262.3 | XP_005257319.1 | 3 | 192 | |||
XM_011524697.2 | XP_011522999.1 | 4 | 234 | |||
GenBank mRNA | AF121861.1 | - | 3 | 384 | ||
AK023932.1 | - | 4 | 451 | |||
AK091852.1 | - | 4 | 349 | |||
AK296569.1 | - | 4 | 433 | |||
AK298551.1 | - | 3 | 187 | |||
AK316374.1 | - | 3 | 184 | |||
BC000768.2 | - | 3 | 181 | 2963905 | ||
BC103721.1 | - | 3 | 170 | 6162638 | ||
BT006723.1 | - | 2 | 97 |