Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130071.1 | NP_001123543.1 | 7 | 1585 | ||
NM_001130072.1 | NP_001123544.1 | 7 | 1122 | |||
NM_013333.3 | NP_037465.2 | 6 | 1047 | |||
XM_005258829.2 | XP_005258886.1 | 7 | 1131 | |||
XM_005258830.2 | XP_005258887.1 | 7 | 1122 | |||
XM_011526880.1 | XP_011525182.1 | 7 | 1131 | |||
XM_011526881.1 | XP_011525183.1 | 6 | 1056 | |||
XM_017026722.1 | XP_016882211.1 | 6 | 1056 | |||
XM_017026723.1 | XP_016882212.1 | 9 | 1173 | |||
XM_017026724.1 | XP_016882213.1 | 9 | 1111 | |||
XM_017026725.1 | XP_016882214.1 | 8 | 1098 | |||
GenBank mRNA | AF073727.1 | - | 7 | 1584 | ||
AK022454.1 | - | 7 | 1091 | |||
AK300545.1 | - | 8 | 995 | |||
AK314690.1 | - | 7 | 1092 | |||
BC044651.1 | - | 6 | 997 | 5244118 |