Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013339.3 | NP_037471.2 | 4 | 525 | ||
GenBank mRNA | AF063604.2 | - | 4 | 480 | ||
AF102851.1 | - | 4 | 458 | |||
AK022700.1 | - | 4 | 463 | |||
AU280221.1 | - | 4 | 488 | |||
BC001253.1 | - | 4 | 457 | 3452687 | ||
BI819904.1 | - | 4 | 527 |