Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013339.3 | NP_037471.2 | 4, 5 | 558 | ||
GenBank mRNA | AF063604.2 | - | 4, 5 | 513 | ||
AF102851.1 | - | 4, 5 | 491 | |||
AK022700.1 | - | 4, 5 | 496 | |||
AU280221.1 | - | 4, 5 | 521 | |||
BC001253.1 | - | 4, 5 | 490 | 3452687 | ||
BI819904.1 | - | 4, 5 | 560 |