Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PURG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015508.2 | NP_001015508.1 | 1 | 1761 | ||
NM_001323311.1 | NP_001310240.1 | 2 | 951 | |||
NM_001323312.1 | NP_001310241.1 | 2 | 951 | |||
NM_013357.2 | NP_037489.1 | 1 | 847 | |||
GenBank mRNA | AF195513.2 | - | 1 | 1761 | ||
AY077841.1 | - | 1 | 1761 | |||
BC106708.2 | - | 1 | 847 | 40005076 |