Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321357.1 | NP_001308286.1 | 19 | 2356 | ||
NM_001321358.1 | NP_001308287.1 | 19 | 1753 | |||
NM_001321359.1 | NP_001308288.1 | 19 | 1748 | |||
NM_001321361.1 | NP_001308290.1 | 19 | 1750 | |||
NM_001321362.1 | NP_001308291.1 | 18 | 1726 | |||
NM_001321363.1 | NP_001308292.1 | 18 | 1633 | |||
NM_013392.3 | NP_037524.1 | 18 | 1724 | |||
GenBank mRNA | AF113249.1 | - | 18 | 1636 | ||
AK001946.1 | - | 18 | 1614 | |||
AK027538.1 | - | 18 | 1632 | |||
AK092860.1 | - | 11 | 2034 | |||
AK122664.1 | - | 19 | 2356 | |||
AK130619.1 | - | 11 | 768 | |||
AK223136.1 | - | 18 | 1620 | |||
AK301345.1 | - | 19 | 1567 | |||
AL136682.1 | - | 18 | 1611 | |||
BC001221.1 | - | 18 | 1638 | 3534543 | ||
BQ225378.1 | - | 4 | 173 |